BCECF (2′,7′ –bis-(Carboxyethyl)-5-(and-6)-carboxyfluorescein) ≥95%

Supplier: ADIPOGEN CORP MS

Synonyms: BCEF acid

CDX-C0002-M001 CDX-C0002-M005 CDX-C0002-M025
102987-506EA 88.62 USD
102987-506 102987-508 102987-510
BCECF (2′,7′ –bis-(Carboxyethyl)-5-(and-6)-carboxyfluorescein) ≥95%
BCECF (2′,7′ –bis-(Carboxyethyl)-5-(and-6)-carboxyfluorescein)

Fluorescent indicator for measurement of cytoplasmic pH in lymphocytes.

Formula: C₂₇H₂₀O₁₁
MW: 520.44 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00036970
CAS Number: 85138-49-4

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Yellow to red solid.
Solubility Soluble in DMSO or dimethylformamide.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR