N-Butanoyl-DL-homoserine lactone ≥97% (by GC)

Supplier: ADIPOGEN CORP MS

Synonyms: BHL, N-(2-Oxotetrahydrofuran-3-yl)butyramide

CDX-B0280-M005 CDX-B0280-M025
102987-502EA 96.92 USD
102987-502 102987-504
N-Butanoyl-DL-homoserine lactone ≥97% (by GC)
N-Butanoyl-DL-homoserine lactone

N-Butyryl-DL-homoserine lactone is a small diffusible signaling molecule and is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones (AHL) are involved in quorum sensing, controlling gene expression, and cellular metabolism. The diverse applications of this kind of molecule include regulation of virulence in general, infection prevention, and formation of biofilms.

Formula: C₈H₁₃NO₃
MW: 171.2 g/mol
Storage Temperature: Freezer
MDL Number: MFCD01862909
CAS Number: 98426-48-3

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (GC)
Appearance/Color White solid.
Solubility Soluble in chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR