N-(3-Bromopropyl)-7-nitro-2,1,3-benzoxadiazol-4-amine ≥98%

Supplier: ADIPOGEN CORP MS

Synonyms: N-(3-Bromopropyl)-7-nitrobenzo[c][1,2,5]oxadiazol-4-amine

CDX-B0268-M100 CDX-B0268-M500
102987-490EA 368.31 USD
102987-490 102987-492
N-(3-Bromopropyl)-7-nitro-2,1,3-benzoxadiazol-4-amine ≥98%
N-(3-Bromopropyl)-7-nitro-2,1,3-benzoxadiazol-4-amine

Building block for synthesis. Used for the preparation of fluorescent probes.

Formula: C₉H₉BrN₄O₃
Storage Temperature: Freezer
CAS Number: 1246965-49-0

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color Brown powder.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR