2-Bromoacrolein ≥95% (by GC)
Supplier: ADIPOGEN CORP MS
|
Danger
|
CDX-B0251-M500
CDX-B0251-G002
102987-482EA
318.46
USD
102987-482
102987-484
2-Bromoacrolein ≥95% (by GC)
2-Bromoacrolein
Building block for synthesis.
Formula:
C₃H₃BrO MW: 134.96 g/mol Storage Temperature: Freezer |
CAS Number:
14925-39-4 UN: 2927 ADR: 6.1 (8),II |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Liquid. |
Solubility | Soluble in chloroform. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...