Bis(4-methylphenyl)iodonium hexafluorophosphate ≥98%

Supplier: ADIPOGEN CORP MS
Danger

Synonyms: Bis(p-tolyl)iodonium hexafluorophosphate

CDX-B0247-G010 CDX-B0247-G125
102989-434EA 234 USD
102989-434 102989-436
Bis(4-methylphenyl)iodonium hexafluorophosphate ≥98%
Bis(4-methylphenyl)iodonium hexafluorophosphate

Building block for synthesis. Reagent used for electrophilic phenylation, e.g. of malonates and dithiocarboxylates.

Formula: C₁₄H₁₄F₆IP
Storage Temperature: Refrigerator
CAS Number: 60565-88-0
UN: 1759
ADR: 8,II

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (NMR)
Appearance/Color White crystalline solid.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR