Furanone C30 ≥97% (by GC)

Supplier: ADIPOGEN CORP MS

Synonyms: (Z)-4-Bromo-5-(brommethylen)-2(5H)-furanone

CDX-B0220-M010 CDX-B0220-M025 CDX-B0220-M100
102989-414EA 131.54 USD
102989-414 102989-416 102989-418
Furanone C30 ≥97% (by GC)
Furanone C30

Quorum sensing inhibitor. Interfers with the actions of acyl-HSL molecules and inhibits gene expression regulated by quorum-sensing systems.

Formula: C₅H₂Br₂O₂
MW: 253.88 g/mol
Storage Temperature: Freezer
MDL Number: MFCD16875408
CAS Number: 247167-54-0

Order Now

Specification Test Results

Source/Host Synthetic
Purity 97% (GC)
Appearance/Color Crystalline.
Solubility Soluble in ether.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR