Nickamine 97
Supplier: ADIPOGEN CORP MS
Synonyms:
Bis[(2-dimethylamino)phenyl]amine nickel(II) chloride
CDX-B0150-M100
CDX-B0150-M500
102989-396EA
157.85
USD
102989-396
102989-398
Nickamine 97
Nickamine
Reagent used as a catalyst for cross-coupling reactions between alkyl halides and carbon nucleophiles. Catalyst for alkyl-alkyl Kumada coupling of secondary alkyl halides, direct alkylation of heterocyclic C-H bonds, Sonogashira coupling of nonactivated alkyl halides, Kumada-Corriu-Tamao coupling of nonactivated alkyl halides with aryl and heteroaryl nucleophiles. Reactant of pincer NN2 ligand leading to selective carbon-carbon bond formation.
Formula:
C₁₆H₂₀ClN₃Ni MW: 348.5 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD17015253 CAS Number: 1033772-47-2 |
Specification Test Results
Source/Host | Synthetic |
Purity |
|
Appearance/Color | Grey powder. |
Solubility | Soluble in DMSO, THF, 1,4-dioxane, chloroform, dichloromethane, DMA and toluene. Insoluble in alkanes and water. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...