(1S)-(+)-3-Bromocamphor-10-sulfonic acid hydrate ≥98% (by TLC)

Supplier: ADIPOGEN CORP MS
Danger

CDX-B0135-M250 CDX-B0135-G001
102989-388EA 78.92 USD
102989-388 102989-390
(1S)-(+)-3-Bromocamphor-10-sulfonic acid hydrate ≥98% (by TLC)
(1S)-(+)-3-Bromocamphor-10-sulfonic acid hydrate

Chiral reagent used for the resolution of bases. Building block for synthesis.

Formula: C₁₀H₁₅BrO₄S·xH₂O
MW: 311.19 g/mol (anhy)
Storage Temperature: Freezer
MDL Number: MFCD00167444
CAS Number: 206860-46-0
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Source/Host Synthetic
Purity 98% (TLC )
Appearance/Color Beige crystalline.
Solubility Soluble in methanol or water.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR