4-(Bromomethyl)phenyl isothiocyanate ≥98%

Supplier: ADIPOGEN CORP MS
Danger

Synonyms: 1-(Bromomethyl)-4-isothiocyanatobenzene

CDX-B0091-M025 CDX-B0091-M100
102989-354EA 62.31 USD
102989-354 102989-356
4-(Bromomethyl)phenyl isothiocyanate ≥98%
4-(Bromomethyl)phenyl isothiocyanate

Hetero-bifunctional linker specially for the quaternisation of pyridyl-nitrogen in order to introduce an amine or thiol reactive group. Synthetic building block for the preparation of fluorescent labels.

Formula: C₈H₆BrNS
Storage Temperature: Freezer
MDL Number: MFCD08702661
CAS Number: 155863-32-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color White to light brown.
Solubility Soluble in toluene.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR