2-(2-Benzofuranyl)-3-hydroxychromone ≥95% (by HPLC)

Supplier: ADIPOGEN CORP MS

CDX-B0069-M020 CDX-B0069-M250
102989-350EA 318.46 USD
102989-350 102989-352
2-(2-Benzofuranyl)-3-hydroxychromone ≥95% (by HPLC)
2-(2-Benzofuranyl)-3-hydroxychromone

Building block for synthesis.

Formula: C₁₇H₁₀O₄
Storage Temperature: Freezer
CAS Number: 67261-94-3

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPLC)
Appearance/Color Solid.
Solubility Soluble in DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR