BTBCT ≥95% (by HPCE)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
4,4″-Bis(4,4,4-trifluoro-1,3-dioxobutyl)-o-terphenyl-4′-sulphonyl chloride
CDX-B0062-M005
CDX-B0062-M050
CDX-B0062-M250
102989-344EA
88.62
USD
102989-344
102989-346
102989-348
BTBCT ≥95% (by HPCE)
BTBCT
Suitable for the fluorometric detection of EU3+.
Formula:
C₂₆H₁₅ClF₆O₆S MW: 604.9 g/mol Storage Temperature: Freezer |
CAS Number:
525560-81-0 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...