8-Butyryloxy-pyren-1,3,6-trisulfonic acid ≥95% (by HPLC)
Supplier: ADIPOGEN CORP MS
|
Warning
|
CDX-B0025-M050
CDX-B0025-M250
102989-318EA
257.54
USD
102989-318
102989-320
8-Butyryloxy-pyren-1,3,6-trisulfonic acid ≥95% (by HPLC)
8-Butyryloxy-pyren-1,3,6-trisulfonic acid
Highly water soluble enzyme substrate for the fluorimetric assay of esterases at longwave excitation and emission wavelengths.
Formula:
C₂₀H₁₃Na₃O₁₁S₃ Storage Temperature: Freezer |
CAS Number:
115787-82-1 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Solid. |
Solubility | Soluble in DMSO, DMF water or methanol. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...