MPAC-Br ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 3-[4-(Bromomethyl)phenyl]7-(diethylamino)-coumarin

CDX-B0018-M010 CDX-B0018-M050 CDX-B0018-M250
102989-312EA 72 USD
102989-312 102989-314 102989-316
MPAC-Br ≥98% (by NMR)
MPAC-Br

HPLC-precolumn-derivatization reagent for carboxylic acids. Compared to classical BMC it emits at longer wavelength and has stronger fluorescence.

Formula: C₂₀H₂₀BrNO₂
MW: 386.28 g/mol
Storage Temperature: Freezer
CAS Number: 177093-58-2

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (NMR)
Appearance/Color Solid.
Solubility Soluble in DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR