N1-Acetylsulfamethoxazol ≥98%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: Acetylsulphisomezole

CDX-A0305-M025 CDX-A0305-M100
102989-292EA 202.15 USD
102989-292 102989-294
N1-Acetylsulfamethoxazol ≥98%
N1-Acetylsulfamethoxazol

Antibacterial pro-drug of sulfamethoxazole.

Formula: C₁₂H₁₃N₃O₄S
MW: 295.31 g/mol
Storage Temperature: Freezer
CAS Number: 18607-98-2

Order Now

Specification Test Results

Source/Host Synthetic
Purity 98% (HPLC)
Appearance/Color White powder.
Solubility Soluble in ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR