S-(5')-Adenosyl-3-thiopropylamine ≥98%
Supplier: ADIPOGEN CORP MS
|
Danger
|
Synonyms:
dc-SAH
CDX-A0210-M010
CDX-A0210-M050
102989-274EA
157.85
USD
102989-274
102989-276
S-(5')-Adenosyl-3-thiopropylamine ≥98%
S-(5')-Adenosyl-3-thiopropylamine
This decarboxylated S-Adenosyl-L-methionine is an important metabolite in polyamine biosynthesis, acting as aminopropyl group donor for propylamine transferases such as spermine synthase and spermidine synthase.
Formula:
C₁₃H₂₀N₆O₃S MW: 340.4 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD17215932 CAS Number: 53186-57-5 UN: 2811 ADR: 6.1 II |
Specification Test Results
Source/Host | Synthetic |
Purity |
|
Appearance/Color | White powder. |
Solubility | Soluble in chloroform. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...