A2B2-Ionophore ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

CDX-A0209-M025 CDX-A0209-M250
102989-270EA 257.54 USD
102989-270 102989-272
A2B2-Ionophore ≥98% (by HPLC)
A2B2-Ionophore

Analytical/Chromatography reagent. Ion sensor compound. Ionophore potentiometric for ISE.

Formula: C₄₄H₃₆N₄O₄
Storage Temperature: Freezer
CAS Number: 1253421-84-9

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color Solid.
Solubility Soluble in acetonitrile.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR