9,10-Anthracenediyl-bis(methylene)dimalonic acid ≥90% (by HPCE)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
2,2'-(Anthracene-9,10-diylbis(methylene))dimalonic acid
CDX-A0183-M050
CDX-A0183-M200
CDX-A0183-G001
102989-240EA
113.54
USD
102989-240
102989-242
102989-244
9,10-Anthracenediyl-bis(methylene)dimalonic acid ≥90% (by HPCE)
9,10-Anthracenediyl-bis(methylene)dimalonic acid
Reagent for the assay of O2 with better characteristics than 9,10-anthracenediyl-bis-dipropionic acid.
Formula:
C₂₂H₁₈O₈ MW: 410.37 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD03095464 CAS Number: 307554-62-7 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | Light brown powder. |
Solubility | Soluble in water or DMSO. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...