3,5-Dimethyl-NAPQI ≥95% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: NA-3,5-Bqi

CDX-A0163-M010 CDX-A0163-M025
102989-212EA 333.69 USD
102989-212 102989-214
3,5-Dimethyl-NAPQI ≥95% (by HPLC)
3,5-Dimethyl-NAPQI

Metabolite. Oxidizing analog of N-acetyl-p-benzoquinone imine (NAPQI), a toxic metabolite of acetaminophen (paracetamol). Primarily oxidizes protein thiols. Shows cytotoxic properties, through disruption of intracellular Ca2+ homeostasis. Compound can be used as analytical reference material.

Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00078877
CAS Number: 74827-85-3

Order Now

Specification Test Results

Source/Host Synthetic
Purity 95.0% (HPLC)
Appearance/Color Orange powder.
Solubility Soluble in chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR