S-BAIBA ≥99%

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: (S)-b-aminoisobutyric acid, L-3-Amino-isobutyric acid, (S)-3-Amino-2-methylpropanoic acid

CDX-A0147-M100 CDX-A0147-M500
102989-192EA 263.08 USD
102989-192 102989-194
S-BAIBA ≥99%
S-BAIBA

Beta-Aminoisobutyric acid is a non-protein amino acid originating from the catabolism of thymine and valine. beta-Aminoisobutyric acid occurs in two isomeric forms and both enantiomers of beta-Aminoisobutyric acid can be detected in human urine and plasma. In plasma, the S-enantiomer is the predominant type due to active renal reabsorption. In contrast, urine almost exclusively contains the R-enantiomer of beta-Aminoisobutyric acid, which is eliminated both by filtration and tubular secretion. The S-enantiomer of beta-Aminoisobutyric acid is predominantly derived from the catabolism of valine, the R-enantiomer is the product of the catabolism of the pyrimidine bases uracil and thymine by the enzyme dihydropyrimidine dehydrogenase (DPD), in what constitutes the first step of the pyrimidine degradation pathway. Transient high levels of beta-Aminoisobutyric acid have been observed under a variety of pathological conditions such as lead poisoning, starvation, in total body irradiation and in a number of malignancies. Recently R-/S-enantiomer mixtures have been shown to be browning inducer of white adipose tissue.

Formula: C₄H₉NO₂
Storage Temperature: Freezer
MDL Number: MFCD07372875
CAS Number: 4249-19-8

Order Now

Specification Test Results

Source/Host Synthetic
Purity 99% (TLC)
Appearance/Color White powder.
Solubility Soluble in methanol or water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR