AQC ≥95% (by NMR)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
6-Aminoquinoline N-succinimidyl ester, 2,5-Dioxo-1-pyrrolidinyl 6-Quinolylcarbamate
CDX-A0057-M050
CDX-A0057-M250
102989-166EA
530.31
USD
102989-166
102989-168
AQC ≥95% (by NMR)
AQC
Suitable for amino acid or protein sequence analysis by HPLC with fluorescence detection.
Formula:
C₁₄H₁₁N₃O₄ MW: 285.25 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD01940654 CAS Number: 148757-94-2 |
Specification Test Results
Source/Host | Synthetic. |
Purity |
|
Appearance/Color | White crystalline powder. |
Solubility | Soluble in DMSO or DMF. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...