5-Aminoeosin ≥97% (by NMR)

Supplier: ADIPOGEN CORP MS

CDX-A0055-M010 CDX-A0055-M025
102989-162EA 175.85 USD
102989-162 102989-164
5-Aminoeosin ≥97% (by NMR)
5-Aminoeosin

Fluorescent marker used extensively in microscopy studies. Can be used in photoluminometric immunoassays for the formation of conjugates with immunoreactants by fluorescent energy transfer.

Formula: C₂₀H₉Br₄NO₅
Storage Temperature: Refrigerator
MDL Number: MFCD00467513
CAS Number: 75900-75-3

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 97% (NMR)
Appearance/Color Red crystalline powder.
Solubility Soluble in chloroform.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR