6-(4-Acetamido-1,8-naphthalamido) hexanoic acid ≥95%

Supplier: ADIPOGEN CORP MS

CDX-A0038-M010 CDX-A0038-M050
102989-152EA 96.92 USD
102989-152 102989-154
6-(4-Acetamido-1,8-naphthalamido) hexanoic acid ≥95%
6-(4-Acetamido-1,8-naphthalamido) hexanoic acid

The highly fluorescent label 4-acetylamino-1,8-naphthalimide-N-caproic acid has two fused benzene rings containing a closed anhydride ring and therefore is a flat and planar molecule which can easily intercalate or act as a groove binder with DNA/RNA sequences. For the study of in situ hydridization with complementary synthetic oligonucleotides (ODNs), tagging with non-radioactive labels has gained importance, not only for medical diagnostics or clinical genetics, but also detection and isolation of DNA for research purposes. The pentamethylene chain in 6 acts as a linker molecule and keeps the target at a reasonable distance to avoid direct interaction with oligonucleotides.

Formula: C₂₀H₂₀N₂O₅
Storage Temperature: Freezer
CAS Number: 172227-59-7

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 95% (HPCE)
Appearance/Color Solid.
Solubility Soluble in water or DMSO.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR