AMQI ≥98% (by HPCE)

Supplier: ADIPOGEN CORP MS

Synonyms: 7-Acetoxy-1-methylquinolinium iodide

CDX-A0036-M100 CDX-A0036-G001
102989-148EA 210.46 USD
102989-148 102989-150
AMQI ≥98% (by HPCE)
AMQI

Fluorogenic substrate for cholinesterase. Non-fluorescent 7-acetoxy-1-methyl quinolinium iodide is hydrolysed to the highly fluorescent 7-hydroxy-1-methyl quinolinium iodide.

Formula: C₁₂H₁₂INO₂
MW: 329.13 g/mol
Storage Temperature: Freezer
CAS Number: 7270-83-9

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPCE)
Appearance/Color Solid.
Solubility Soluble in DMSO or DMF.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR