DAF-FM ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: 4-Amino-5-methylamino- 2',7'-difluorofluorescein

CDX-A0023-M001 CDX-A0023-M005
102989-140EA 508.15 USD
102989-140 102989-142
DAF-FM ≥98% (by HPLC)
DAF-FM

DAF-FM is an important reagent for quantitating low concentrations of nitric oxide in solution. This compound is essentially nonfluorescent until it reacts with NO to form a fluorescent benzotrizole. With excitation/emission maxima of 495/515 nm, DAF-FM can be detected using any instrument that can be used to detect fluorescein.

Formula: C₂₁H₁₄F₂N₂O₅
MW: 412.34 g/mol
Storage Temperature: Freezer
CAS Number: 254109-20-1

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color Orange solid.
Solubility Soluble in DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR