10-Acetyl-3,7-dihydroxyphenoxazine ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

CDX-A0022-M025 CDX-A0022-M200
102989-136EA 333.69 USD
102989-136 102989-138
10-Acetyl-3,7-dihydroxyphenoxazine ≥98% (by HPLC)
10-Acetyl-3,7-dihydroxyphenoxazine

10-acetyl-3,7-dihydroxyphenoxazin (Amplex Red) is a non-fluorescent, highly sensitive and stable probe for H2O2. Amplex Red is an applicable as a fluorogenic substrate for peroxidase. In the presence of horseradish peroxidase (HRP), the Amplex Red reagent reacts in a 1:1 stechiometry with H2O2 to produce highly fluorescent resorufin. Because H2O2 is produced in many different enzymatic reactions, the Amplex Red reagent allows researchers to detect the activity of many different enzymes. 10-acetyl-3,7-dihydroxyphenoxazin has absorption and fluorescence emission maxima of approximately 563 nm and 587 nm, respectively, and because its extinction coefficient is high (54,000 cm-1M-1), the assay can be performed either fluorometrically or spectrophotometrically.

Formula: C₁₄H₁₁NO₄
Storage Temperature: Freezer
MDL Number: MFCD00467861
CAS Number: 119171-73-2

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98% (HPLC)
Appearance/Color Solid.
Solubility Soluble in DMSO or DMF.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR