AMCA-H N-succinimidyl ester ≥90% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: Succinimidyl-7-amino-4-methylcoumarin-3-acetate, N-Succinimidyl 7-amino-4-methyl-3-coumarinylacetate, NHS-AMCA, 7-Amino-4-methyl-3-coumarinacetic acid N-succinimidyl ester, 7-(Dimethylamino)coumarin-4-acetic acid-NHS, AMCA-H-N-S-ester, 7-Amino-4-methyl-3coumarinylacetyl

CDX-A0010-M025 CDX-A0010-M100
102989-116EA 139.85 USD
102989-116 102989-118
AMCA-H N-succinimidyl ester ≥90% (by HPLC)
AMCA-H-N-succinimidyl ester

AMCA-H NHS ester reacts with primary amine groups on antibodies, proteins, peptides and other biomolecules at pH 7.0-9.0. Suitable for labeling lysyl residues in proteins (Ex/Em: 353/455nm).

Formula: C₁₆H₁₄N₂O₆
MW: 330.29 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00036170
CAS Number: 113721-87-2

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 90% (HPLC)
Appearance/Color Yellow powder.
Solubility Soluble in DMSO or DMF.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR