Luteoreticulin ≥98% (by HPLC, NMR)

Supplier: ADIPOGEN CORP MS

Synonyms: Griseulin

BVT-0457-C500 BVT-0457-M001
102989-080EA 193.85 USD
102989-080 102989-082
Luteoreticulin ≥98% (by HPLC, NMR)
Luteoreticulin

Nitrophenyl pyranone derivative related to aureothin . Insecticidal and nematocidal compound. Metabolites within this product class show pronounced antitumor and immunosuppressive activity.

Formula: C₁₉H₁₉NO₅
MW: 341.4 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14635388
CAS Number: 22388-89-2

Order Now

Specification Test Results

Source/Host Isolated from Streptomyces thioluteus.
Purity 98% (NMR, HPLC)
Appearance/Color Yellow solid.
Solubility Soluble in DMSO, acetone or chloroform.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR