Butyrolactone I ≥98% (by HPLC, NMR)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Cell permeable, potent and selective cyclin-dependent kinase CDK-1 (CDC2), -2 and -5 inhibitor. Apoptosis inducer. Antitumor agent. Inhibits cell cycle progression at the G1/S and G2/M transitions. Shown to prevent the phosphorylation of retinoblastoma protein and H1 histone. Important probe for understanding the cellular roles of CDKs. Inhibitor of alpha-glucosidases. Maturation inhibitor of sheep oocytes.
Formula:
C₂₄H₂₄O₇ Storage Temperature: Freezer |
MDL Number:
MFCD03453074 CAS Number: 87414-49-1 |
Specification Test Results
Source/Host | Isolated from Aspergillus terreus. |
Purity |
|
Appearance/Color | White to off-white solid. |
Solubility | Soluble in DMSO, methanol, ethanol or acetone. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...