3-O-Caffeoyl-betulin ≥96% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: Betulin 3-caffeate

BVT-0393-M001 BVT-0393-M005
102988-946EA 110.77 USD
102988-946 102988-948
3-O-Caffeoyl-betulin ≥96% (by HPLC)
3-O-Caffeoyl-betulin

Anti-proliferative. Shows anti-melanoma activity. Sun-block ingredient for cosmetics. Prevents sun burning, premature aging and skin cancer. Antibacterial.

Formula: C₃₉H₅₆O₅
MW: 604.9 g/mol
Storage Temperature: Freezer
CAS Number: 89130-86-9

Order Now

Specification Test Results

Source/Host Isolated from birch bark (Betula alba).
Purity 96% (HPLC)
Appearance/Color Light yellow solid.
Solubility Soluble in methanol, acetone or DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR