Chloroguanabenz acetate ≥95% (by NMR)

Supplier: ADIPOGEN CORP MS
Danger

Synonyms: 1-[(E)-2,3,6-Trichlorophenylmethyleneamino]guanidine

AG-MR-C0036-M001 AG-MR-C0036-M005
102988-870EA 83.08 USD
102988-870 102988-872
Chloroguanabenz acetate ≥95% (by NMR)
Chloroguanabenz acetate

Antiprion agent. Inhibitor of protein aggregation. Specifically reduces accumulation of a pathogenic fragment of Huntingtin in a transiently transfected cellular model of Huntington’s disease (HD). Tool for treating HD and other polyglutamine expansion associated diseases.

Formula: C₈H₇Cl₃N₄·C₂H₄O₂
MW: 325.6 g/mol
Storage Temperature: Freezer
CAS Number: 23113-55-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Purity 95% (NMR)
Appearance/Color White to off-white powder.
Solubility Soluble in DMSO or ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR