6-Amino-8-trifluoromethylphenanthridine ≥99% (by NMR)

Supplier: ADIPOGEN CORP MS
Warning

AG-MR-C0031-M001 AG-MR-C0031-M005 AG-MR-C0031-M025
102988-864EA 83.08 USD
102988-864 102988-866 102988-868
6-Amino-8-trifluoromethylphenanthridine ≥99% (by NMR)
6-Amino-8-trifluoromethylphenanthridine

Potent antiprion agent. Higher activity than 6-Aminophenanthridine . Ribosome-borne protein folding activity (RPFA) inhibitor. Binds to the ribosomal RNA and inhibits specifically the protein folding activity of the ribosome. Inhibitor of protein aggregation.

Formula: C₁₄H₉F₃N₂
Storage Temperature: Freezer
CAS Number: 651055-83-3

Order Now

Specification Test Results

Purity 99% (NMR)
Appearance/Color Beige solid.
Solubility Soluble in DMSO or ethanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR