Me6BIO ≥95% (by NMR)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: (2Z,3E)-6'-Bromo-3-(hydroxyimino)-1'-methyl-[2,3'-biindolinylidene]-2'-one, MeBIO

AG-MR-C0021-M005 AG-MR-C0021-M025
102988-844EA 138.46 USD
102988-844 102988-846
Me6BIO ≥95% (by NMR)
Me6BIO

N-methylated inactive control analog of 6-bromoindirubin-3'-oxime (6BIO) . Displays minimal activity against CDK1/Cyclin B, GSK-3 alpha/beta and CDK5/p25. Aryl hydrocarbon receptor (AhR) agonist; causes redistribution of AhR to the nucleus.

Formula: C₁₇H₁₂BrN₃O₂
MW: 370.2 g/mol
Storage Temperature: Freezer
CAS Number: 667463-95-8

Order Now

Specification Test Results

Purity 95% (NMR)
Appearance/Color Dark red solid.
Solubility Soluble in DMSO or ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR