7BIO ≥99% (by NMR)
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
7'-Bromo-3-(hydroxyimino)-[2,3'-biindolinylidene]-2'-one, 7-Bromoindirubin-3'-oxime
AG-MR-C0020-M010
AG-MR-C0020-M050
102988-840EA
193.85
USD
102988-840
102988-842
7BIO ≥99% (by NMR)
7BIO
Anticancer compound. Caspase-independent, non-apoptotic cell death inducer. ATP-competitive Flt3 inhibitor. Necrosis inducer. Aurora B and C kinase inhibitor. DYRK1A and DYRK 2 (Dual-specificity tyrosine phosphorylation-regulated kinase) inhibitor. Inflammasome activator.
Formula:
C₁₆H₁₀BrN₃O₂ MW: 356.2 g/mol Storage Temperature: Freezer |
CAS Number:
916440-85-2 |
Specification Test Results
Purity |
|
Appearance/Color | Dark red solid. |
Solubility | Soluble in DMSO or ethanol. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...