beta-Mannosylceramide ≥95% (by TLC, NMR)

Supplier: ADIPOGEN CORP MS

Synonyms: beta-ManCer

AG-CR1-3621-C250 AG-CR1-3621-M001
102988-800EA 221.54 USD
102988-800 102988-802
beta-Mannosylceramide ≥95% (by TLC, NMR)
beta-Mannosylceramide

Potent stimulator of invariant natural killer T (iNKT) cells. Induces strong antitumor immunity through a nitric oxide (NO) and TNF-alpha dependent mechansim, different compared to the IFN-gamma dependent mechanism of alpha-GalCer (AG-CN2-0014). Does not induce anergy of iNKT cells.

Formula: C₅₀H₉₉NO₉
MW: 858.3 g/mol
Storage Temperature: Freezer
CAS Number: 1334332-65-8

Order Now

Specification Test Results

Purity 95% (TLC, NMR)
Appearance/Color White to off-white solid.
Solubility Soluble in pyridine or warm DMSO. Insoluble in water.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR