Empagliflozin ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

Synonyms: (2S,3R,4R,5S,6R)-2-(4-Chloro-3-(4-(((S)-tetrahydrofuran-3-yl)oxy)benzyl)phenyl)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol, Jardiance

AG-CR1-3619-M010 AG-CR1-3619-M050
102988-792EA 96.92 USD
102988-792 102988-794
Empagliflozin ≥98% (by HPLC)
Empagliflozin

Antidiabetic agent. Potent and selective sodium glucose co-transporter 2 (SGLT-2) inhibitor. 300-fold selectivity over SGLT-1, 4, 5 and 6. SGLT-2 is found almost exclusively in the proximal tubules of nephronic components in kidneys. Inhibition of SGLT-2 reduces blood glucose by blocking renal glucose reabsorption and thereby increasing urinary glucose excretion (UGE). Shown to preserves beta cell mass and restore glucose homeostasis.

Formula: C₂₃H₂₇ClO₇
MW: 450.9 g/mol
Storage Temperature: Freezer
MDL Number: MFCD22566222
CAS Number: 864070-44-0

Order Now

Specification Test Results

Purity 98% (HPLC)
Appearance/Color White solid.
Solubility Soluble in DMSO. Slightly soluble in water or ethanol.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR