(S)-3-Hydroxybutyric acid ≥98% (by TLC)

Supplier: ADIPOGEN CORP MS
Warning

AG-CR1-3617-M025 AG-CR1-3617-M100
102988-784EA 41.54 USD
102988-784 102988-786
(S)-3-Hydroxybutyric acid ≥98% (by TLC)
(S)-3-Hydroxybutyric acid

Physiologically insignificant stereoisomer of (R)-3-Hydroxybutyric acid, a key metabolite of the ketolytic pathway. Anticonvulsant. Endogenous inhibitor of histone deacetylases (HDACs) 1, 3 and 4. Ligand of free fatty acid receptor 3 (FFAR3; GPR41) and hydroxycarboxylic acid receptor 2 (HCAR2; GPR109B). NLRP3 inflammasome inhibitor. Prevents K+-efflux and reduces ASC oligomerization and speck formation.

Formula: C₄H₈O₃
Storage Temperature: Freezer
MDL Number: MFCD00137685
CAS Number: 6168-83-8

Order Now

Specification Test Results

Purity 98% (TLC)
Appearance/Color White solid.
Solubility Soluble in water, 100% ethanol or methanol.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR