Auranofin ≥98%

Supplier: ADIPOGEN CORP MS
Danger

Synonyms: ((2S,3R,4S,5R,6R)-3,4,5-Triacetoxy-6-(acetoxymethyl)tetrahydro-2H-pyran-2-thio)(triethylphosphine)gold

AG-CR1-3611-M025 AG-CR1-3611-M100
102988-760EA 76.15 USD
102988-760 102988-762
Auranofin ≥98%
Auranofin

Anti-inflammatory and anti-arthritic agent


Purity: >98%
Appearance/Color: White to off-white solid.
Solubility: Soluble in DMSO or 100% ethanol. Insoluble in water.


Represses the activation of the NLRP3/IL-1beta pathway. Inhibitor of 5-lipoxygenase in human neutrophils, IKB kinase (IKK) by modifying Cys-179 of the IKKbeta subunit 5. Potent thioredoxin reductase (TrxR) inhibitor. Induces mitochondrial membrane permeability transition. Anti-cancer compound with anti-proliferative activity. Shown to induce apoptosis and necrosis. STAT3 and telomerase activity inhibitor. Proteasomal deubiquitinase inhibitor. Antibiotic, antiviral and antiparasitic compound. Immunosuppressive.

Formula: C₂₀H₃₆AuO₉PS
MW: 680.4998 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00080759
CAS Number: 34031-32-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR