BLZ-945 ≥95%
Supplier: ADIPOGEN CORP MS
Synonyms:
BLZ945, Sotuletinib, 4-((2-(((1R,2R)-2-Hydroxycyclohexyl)amino)benzo[d]thiazol-6-yl)oxy)-N-methylpicolinamide
AG-CR1-3609-M001
AG-CR1-3609-M005
AG-CR1-3609-M025
102988-748EA
55.38
USD
102988-748
102988-750
102988-752
BLZ-945 ≥95%
BLZ-945
Anticancer compound. Orally active, potent and selective colony stimulating factor 1 receptor (CSF-1R; c-Fms) inhibitor. More than 1000-fold selective against its closest receptor tyrosine kinase homologs c-KIT and platelet-derived growth factor receptor beta (PDGFRbeta). Selective against more than 200 additional kinases.
Formula:
C₂₀H₂₂N₄O₃S MW: 398.5 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD28142668 CAS Number: 953769-46-5 |
Specification Test Results
Purity |
|
Appearance/Color | Pale yellow solid. |
Solubility | Soluble in DMSO. Slightly soluble in ethanol. Insoluble in water. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...