alpha-Mannosylceramide ≥95% (by TLC)

Supplier: ADIPOGEN CORP MS

Synonyms: alpha-ManCer

AG-CR1-3594-C250 AG-CR1-3594-M001
102988-700EA 186.92 USD
102988-700 102988-702
alpha-Mannosylceramide ≥95% (by TLC)
alpha-Mannosylceramide

Potent stimulator of natural killer T (NKT) cells expressing the invariant Valpha19-Jalpha26 T cell receptor alpha chain (Valpha19 NKT cells).

Formula: C₅₀H₉₉NO₉
MW: 858.3 g/mol
Storage Temperature: Freezer
CAS Number: 320610-63-7

Order Now

Specification Test Results

Purity 95% (TLC)
Appearance/Color White to off-white solid.
Solubility Soluble in pyridine or warm DMSO.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR