cGAMP disodium salt ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS

AG-CR1-3588-C100 AG-CR1-3588-C500
102988-670EA 131.54 USD
102988-670 102988-672
cGAMP disodium salt ≥98% (by HPLC)
cGAMP disodium salt

Potent STING ligand. Second messenger in human and mouse cells that triggers interferon production in response to cytosolic DNA. Binding to STING leads to activation of IRF3 (IFN regulatory factor 3) and the induction of interferon-beta. First cyclic nucleotide in metazoa. Plays a role in bacterial chemotaxis and colonization.

Order Now

Specification Test Results

Source/Host Synthetic.
Purity >98% (HPLC)
Appearance/Color Crystalline solid.
Solubility Soluble in water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR