Gardiquimod ≥98%

Supplier: ADIPOGEN CORP MS
Danger

Synonyms: 1-(4-Amino-2-ethylaminomethylimidazo[4,5-c]quinolin-1-yl)-2-methylpropan-2-ol

AG-CR1-3583-M005 AG-CR1-3583-M025
102988-646EA 55.38 USD
102988-646 102988-648
Gardiquimod ≥98%
Gardiquimod

Immune response modifier. Antiviral and antitumor compound Selective ligand for human or mouse Toll-like receptor 7 (TLR7). Induces the activation of NF-kappaB in HEK 293 cells expressing TLR7 at 0.1µg/ml. 10 times more active than Imiquimod . At high concentrations (3µg/ml) slightly activates TLR8. Shows adjuvant activity to increase the effectiveness of vaccines.

Formula: C₁₇H₂₃N₅O
MW: 313.4 g/mol
Storage Temperature: Freezer
CAS Number: 1020412-43-4
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Source/Host Synthetic.
Purity 98%
Appearance/Color White to off-white powder.
Solubility Soluble in DMSO, dioxane, methanol or acetonitrile.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR