D-NMMA monoacetate ≥98%
Supplier: ADIPOGEN CORP MS
AG-CR1-3580-M005
AG-CR1-3580-M025
AG-CR1-3580-M100
102988-632EA
69.23
USD
102988-632
102988-634
102988-636
D-NMMA monoacetate ≥98%
D-NMMA monoacetate
Cell permeable control compound for L-NMMA . Inactive, shows no NOS inhibitor activity. Useful tool to investigate non-specific L-NMMA activity.
Formula:
C₇H₁₆N₄O₂·CH₃COOH Storage Temperature: Freezer |
CAS Number:
137694-75-8 |
Specification Test Results
Purity |
|
Appearance/Color | White solid. |
Solubility | Soluble in water or methanol. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...