7-Azido-4-methylcoumarin 98
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
7-Azido-4-methyl-1-benzopyran-2-one
Highly sensitive and selective fluorogenic H2S probe. The aromatic azide moiety of AzMC is selectively reduced in the presence of H2S, producing the fluorescent 7-amino-4-methylcoumarin (AMC) with a concomitant increase in fluorescence with lambdaex = 365 nm and lambdaem = 450 nm. Photoaffinity labeling probe for the substrate binding site of human sulfotransferase 1A1 (SULT1A1). Probe to monitor the enzymatic production of H2S in vitro and to visualize H2S in living cells. Tool for monitoring the activity of pyridoxal-5'-phosphate (PLP)-dependent enzymes (e.g. cystathionine beta-synthase (CBS), cystathionine gamma-lyase (CGL) and tryptophan synthase (TS)). Tool to identify novel cystathionine beta-synthase (CBS) inhibitors and activators. Suitable for high-throughput screening. Caution: Use of this product with DTT, TCEP and/or biological thiols at concentrations of >25 mM should be avoided for maximum efficiency.
Formula:
C₁₀H₇N₃O₂ MW: 201.2 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD16652405 CAS Number: 95633-27-5 |
Specification Test Results
Purity | >98% (NMR) |
Appearance/Color | White to yellow solid. |
Solubility | Soluble in DMSO. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...