22-NHC (inactive isomer) ≥98%
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
(3beta)-20R-[(3-Methylbutyl)amino]-pregn-5-en-3-ol
AG-CR1-3524-M001
AG-CR1-3524-M005
102988-586EA
55.38
USD
102988-586
102988-588
22-NHC (inactive isomer) ≥98%
22-NHC (inactive isomer)
Inactive control compound for hedgehog signaling inhibitor 22-NHC . HCl http://www.adipogen.com/ag-cr1-3523/22-nhc-hydrochloride.html .
Formula:
C₂₆H₄₅NO Storage Temperature: Freezer |
CAS Number:
40521-33-3 |
Specification Test Results
Purity |
|
Appearance/Color | White solid. |
Solubility | Soluble in DMSO. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...