22-NHC hydrochloride ≥98%
Supplier: ADIPOGEN CORP MS
|
Warning
|
Synonyms:
22-Azacholesterol
AG-CR1-3523-M001
AG-CR1-3523-M005
102988-582EA
76.15
USD
102988-582
102988-584
22-NHC hydrochloride ≥98%
22-NHC hydrochloride
Aza-steroid. Hedgehog signaling inhibitor. Competes with 20(S)-OHC for access to the cysteine-rich domain in Smoothened (Smo). For control compound see 22-NHC (inactive isomer) http://www.adipogen.com/ag-cr1-3524/22-nhc-inactive-isomer.html .
Formula:
C₂₆H₄₅NO·HCl Storage Temperature: Freezer |
CAS Number:
360068-87-7 |
Specification Test Results
Purity |
|
Appearance/Color | White solid. |
Solubility | Soluble in DMSO. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...