Tripolin A ≥98%
Supplier: ADIPOGEN CORP MS
Synonyms:
(3E)-3-[(2,5-Dihydroxyphenyl)methylene]-1,3-dihydro-2H-indol-2-one
AG-CR1-3520-M001
AG-CR1-3520-M005
102988-574EA
110.77
USD
102988-574
102988-576
Tripolin A ≥98%
Tripolin A
Specific non-ATP competitive Aurora A kinase inhibitor. Does not significantly inhibit Aurora B kinase in mammalian cells. Reduces localization of pAurora A on spindle microtubules (MTs), affects centrosome integrity, spindle formation and length and MT dynamics in interphase.
Formula:
C₁₅H₁₁NO₃ MW: 253.3 g/mol Storage Temperature: Freezer |
CAS Number:
1148118-92-6 |
Specification Test Results
Purity |
|
Appearance/Color | Yellow solid. |
Solubility | Soluble in DMSO (at least 10 mg/ml). |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...