NSC697923 ≥98% (by NMR)
Supplier: ADIPOGEN CORP MS
Selective Ub-conjugating enzyme (E2) complex Ubc13-Uev1A inhibitor. Inhibits the formation of the Ubc13~Ub conjugate. NF-kappaB activation inhibitor. Inhibits proliferation and survival of ABC-DLBCL and GCB-DLBCL (diffuse large B cell lymphoma) cells.
Formula:
C₁₁H₉NO₅S Storage Temperature: Freezer |
MDL Number:
MFCD01925132 CAS Number: 343351-67-7 |
Specification Test Results
Purity |
|
Appearance/Color | White solid. |
Solubility | Soluble in DMSO. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...