Compound 112254 hydrochloride ≥98% (by HPLC, NMR), water soluble
Supplier: ADIPOGEN CORP MS
Adiponectin receptor (AdipoR) agonist. Binds to AdipoR1 and AdipoR2 less potent than AdipoRon http://www.adipogen.com/ag-cr1-0154/adiporon.html . Activates 5'-adenosine monophosphate-activated protein kinase (AMPK) in cultured mammalian cells. Improves diabetes, glucose and lipid metabolism and insulin sensitivity in cultured cells and in mice by AdipoR-dependent mechanisms.
Formula:
C₂₆H₃₅N₃O₃·HCl·H₂O Storage Temperature: Freezer |
CAS Number:
949745-75-9 |
Specification Test Results
Purity | |
Appearance/Color | White crystalline solid |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...