Compound 112254 ≥98% (by NMR)

Supplier: ADIPOGEN CORP MS

Synonyms: 4-(tert-Butyl)-N-(3-(4-(4-methoxybenzyl)piperazin-1-yl)-3-oxopropyl)benzamide

AG-CR1-0155-M010 AG-CR1-0155-M050
102988-548EA 110.77 USD
102988-548 102988-550
Compound 112254 ≥98% (by NMR)
Compound 112254

Adiponectin receptor (AdipoR) agonist. Binds to AdipoR1 and AdipoR2 less potent than AdipoRon http://www.adipogen.com/ag-cr1-0154/adiporon.html . Activates 5'-adenosine monophosphate-activated protein kinase (AMPK) in cultured mammalian cells. Improves diabetes, glucose and lipid metabolism and insulin sensitivity in cultured cells and in mice by AdipoR-dependent mechanisms.

Formula: C₂₆H₃₅N₃O₃
MW: 437.6 g/mol
Storage Temperature: Freezer
CAS Number: 949745-75-9

Order Now

Specification Test Results

Purity 98% (NMR)
Appearance/Color White crystalline solid.
Solubility Soluble in DMSO, 100% ethanol, methanol or dichloromethane. Insoluble in water.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR