LY-333,531 hydrochloride ≥98% (by HPLC)

Supplier: ADIPOGEN CORP MS
Warning

AG-CR1-0087-M001 AG-CR1-0087-M005
102988-540EA 96.92 USD
102988-540 102988-542
LY-333,531 hydrochloride ≥98% (by HPLC)
LY-333,531 hydrochloride

Isozyme selective inhibitor of protein kinase Cbeta (PKCbeta). PKCbetaI and PKCbetaII isozyme inhibitor. Amelioriates diabetic retinopathy, diabetic peripheral neuropathy and diabetic nephropathy. Anti-cancer and anti-angiogenic compound. Suppresses glucose-induced adhesion of human monocytes to endothelial cells. Suppresses ERK1/2 and Akt phosphorylation.

Formula: C₂₈H₂₈N₄O₃·HCl
Storage Temperature: Freezer
MDL Number: MFCD00940226
CAS Number: 169939-93-9

Order Now

Specification Test Results

Purity 98% (HPLC)
Appearance/Color Red solid.
Solubility Soluble in DMSO. Slightly soluble in 100% ethanol or water.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR